missing translation for 'onlineSavingsMsg'
Learn More

EVOS™ EVOS™ Stage Plate for Automated Stage, one Nunc T-175 flask

EVOS™ vessel holders and stage plates fit a wide range of vessels, flasks, plates, dishes, and slides.

Brand:  EVOS™ AMEPVH042

Code : NEW

Additional Details : Weight : 0.50000kg

Product Code. 15471074

  • £273.00 / Each
Estimated Shipment: 07-08-2024
to see stock.

Description

Description

Product Features:
  • EVOS™ vessel holders allow a perfect fit of your microscope slide, cell culture flask or dish, or micro-well plate to the stage of the EVOS imaging systems for increased precision in sample alignment.
  • EVOS stage plates hold vessel holders, thereby expanding the number of vessel holders available for the EVOS systems.
  • With a snug and secure fit, yet readily switched, these accessories make imaging samples in a range of vessel holders convenient and easy.
Specifications

Specifications

Imaging System Stage Plate
Stage Plate for Automated Stage, one Nunc™ T-175 flask
1 Nunc T-175 flask
Room Temperature
EVOS
EVOS™ FL Auto Imaging System
259 mm x 204 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.