Learn More
EVOS™ EVOS™ Onstage Master Plate
Allows user to utilize different types of vessels for imaging
Brand:  EVOS™ AMEPVH035
Code : NEW
Additional Details : Weight : 2.00000kg
Product Code. 15399651
  • £404.00 / Each
Estimated Shipment: 19-08-2024
to see stock.
Add to basket
Description
Description
  • Part of EVOS Onstage Incubator and holds EVOS Onstage Vessel Holder
  • Is an environmental chamber designed specifically for use with EVOS FL Auto Imaging System
  • Accommodates various types of cell culture vessels
  • Provides flexibility to extended time-lapse imaging experiments under precisely controlled environmental conditions
Specifications
Specifications
Master Plate
EVOS™ FL Auto Imaging System
1 Ea.
EVOS
Onstage Vessel