Learn More
Invitrogen™ EVOS™ M3000 Attachable Mechanical Stage
Description
The EVOS M3000 Mechanical Stage is an optional addition to the universal, fixed stage provided with the EVOS M3000 Imaging System. This mechanical stage has X-Y axis controls and a vessel holder framework that accommodates our broad range of vessel holders.
The EVOS M3000 Mechanical Stage is an optional addition to the universal, fixed stage provided with the EVOS M3000 Imaging System. This mechanical stage has X-Y axis controls and a vessel holder framework that accommodates our broad range of vessel holders. Larger vessels, such as T-75 flasks or multi-well plates, fit directly onto the stage. Both vessel holders and large vessels are held securely in place by a stage clip during operation. The spring-loaded clip allows fast and easy vessel changes, such as during operations inside a hood or during processing of multiple samples.
Specifications
Specifications
| Type | Mechanical Stage |
| For Use With (Equipment) | EVOS M3000 Imaging System |
| Quantity | 1 stage |