Learn More
EVOS™ EVOS™ 40X Objective, fluorite, LWD
Components manufactured to ensure publication-quality images
Brand: EVOS™ AMEP4625
Code : NEW
Additional Details : Weight : 0.01000kg
Description
RMS-threaded objectives with 45mm parfocal distance
Specifications
Objective Lens | |
EVOS™ FL Color Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System | |
Plan Fluorite | |
0.65 |
EVOS | |
Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates | |
40X | |
1 Ea. |
For Research Use Only. Not for use in diagnostic procedures.