missing translation for 'onlineSavingsMsg'
Learn More

EVOS™ EVOS™ 10X Objective, fluorite, LWD

Product Code. 11927275
Click to view available options
Quantity:
1 each
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11927275

Brand: EVOS™ AMEP4623

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Components manufactured to ensure publication-quality images

RMS-threaded objectives with 45mm parfocal distance

TRUSTED_SUSTAINABILITY

Specifications

Type EVOS LWD Fluorite Objective
Product Line EVOS
For Use With (Equipment) EVOS™ FL Color Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Includes Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates
Lens Type Plan Fluorite
Magnification 10X
Numerical Aperture 0.30
Quantity 1 each

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.