Learn More
EVOS™ EVOS™ 4X Objective, fluorite, LWD, phase-contrast
Description
RMS-threaded objectives with 45mm parfocal distance
Specifications
Specifications
| Type | EVOS LWD Fluorite Phase Contrast Objective |
| Product Line | EVOS |
| For Use With (Equipment) | EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System |
| Lens Type | Fluorite |
| Magnification | 4X |
| Numerical Aperture | 0.13 |
| Quantity | 1 each |