Learn More
Invitrogen™ EVOS™ Light Cube, YFP
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
- Ideal for use with EVOS FL Color Imaging System, EVOS FL Auto Imaging System or EVOS FL Imaging Systems
- Each cube contains an LED, condensing optics, and hard-coated filters
- Continuous illumination enables consistent results 50,000+ hour bulb lifetime that affords lower operating costs
- Adjustable light intensity that helps reduce photobleaching User changeable and automatically recognized by the system
Specifications
Specifications
| Type | YFP Fluorescent Light Cube |
| Product Line | EVOS |
| For Use With (Equipment) | Countess™ II FL Automated Cell Counter, EVOS™ M5000 Imaging System, EVOS™ M7000 Imaging System |
| Includes | LED, Condensing Optics, Hard-coated Filters |
| Wavelength | Ex: 500/24 Em: 542/27 |
| Quantity | 1 Ea. |