Learn More
EVOS™ EVOS™ Condenser Slider, Meniscus A (clear liquids)
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
- Used to improve illumination under certain conditions
- Due to variances in sample size, color and thickness, actual slider use may differ from suggestions
- Meniscus A slider is intended for brightfield applications at 2x magnification with low-volume samples in multi-well plates and with clear liquids
- Meniscus B slider is intended for brightfield applications at 2x magnification with high-volume samples in multi-well plates and with semi-opaque liquids
Specifications
Specifications
| Type | Meniscus A Condenser Slider |
| Product Line | EVOS |
| For Use With (Equipment) | EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System |
| Includes | Diffuser Slider, Pinhole Slider, Meniscus Sliders |
| Quantity | 1 Ea. |