missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EVORF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£281.00 - £417.00
Specifications
| Antigen | EVORF |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18452951
|
Novus Biologicals
NBP1-93723-25ul |
25 μL |
£281.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18071787
|
Novus Biologicals
NBP1-93723 |
0.1 mL |
£417.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EVORF Polyclonal specifically detects EVORF in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| EVORF | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 29105 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GFLSILYSIGSKPLQIWDKKVRNGHIKRITDNDIQSLVLEIEGTNVSTTYITCPADPKKTLGIKLPFLVMIIKNLKKYFTFEVQVLDDKNVRRRFRASNYQSTTRVKPFICTMPMRLDDGWNQIQFNLL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Human | |
| C16orf80, chromosome 16 open reading frame 80, fSAP23, GTL3 | |
| C16orf80 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title