Learn More
Safety First Aid Evolution Plus Hydrofluoric Acid Burn Kit
Product Code. 31759414
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Each
Product Code. Sterility Unit Size
31759414 Sterile Each
1 options
Product Code. 31759414 Supplier Safety First Aid Supplier No. K384
  • ONLINE EXCLUSIVE PRICE
    £50.71
    £49.95 / Pack of 1
    Save £0.76
    1% Off
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
The kit provides supplies for dealing with hydrofluoric acid skin burns. The kit includes calcium gluconate gel helping to relieve pain and heal the burn, reducing the risk of hypocalcaemia.
  • The kit is supplied in an Evolution case with a quick release wall mounting bracket, meaning the kit can be safely stored on the wall in a visible location and quickly removed to be taken to a casualty.
  • Kit Contents:
    • 4x HypaCover Low Adherent Absorbent Dressings, 10 x 10 cm
    • 4x HypaBand Conforming Bandages, 7.5 cm x 4 m
    • 4x HypaTouch Nitrile Gloves
    • 2x C-gel Calcium Gluconate, 40 g
Specifications
Product Type First Aid Kit
Material Plastic
Sterility Sterile