Learn More
Safety First Aid Evolution Plus First Aid Station, Large
Product Code. 31759434
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Each
Product Code. Sterility Unit Size
31759434 Sterile Each
1 options
Product Code. 31759434 Supplier Safety First Aid Supplier No. FAS01
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Evolution First Aid Station is the ultimate first aid solution for any workplace. It is combining a British Standard compliant first aid kit in a durable Evolution case with dispensers for Salvequick plasters, wound wipes and eye wash pods.
  • Every item is wall-mounted to form a single panel of first aid essentials alongside crucial first aid guidance for emergencies.
  • The case comes with a wall bracket featuring an innovative one-handed quick-release catch.
  • The kits contents comply fully with the British Standard minimum content for workplace first aid kits.
  • 2 tamper-proof studs are provided to deter people from opening the kit unnecessarily.
  • This also greatly simplifies regular first aid inspections (as recommended by the HSE) as you can see at a glance whether the kit has been opened and contents likely removed.
  • To ensure the kit won’t be opened for minor incidents, the Evolution First Aid Station also includes three handy dispensers to give quick access to popular items without depleting the first aid box.
  • Plasters are the most commonly used item and the most likely to be taken for home use.
  • The Salvequick pilfer-proof plaster dispenser prevents this, as it’s only possible to remove one plaster at a time, and removing the plaster immediately exposes the adhesive.
  • Plasters can easily be applied with one hand.
  • The dispenser is locked to prevent the plaster packet being removed.
  • It contains high-quality allergenic fabric and washproof plasters in two sizes.
  • In addition to the plaster dispenser, an eye wash dispenser contains 10 20 mL sterile saline eye wash pods, for rinsing irritants like dust or small insects out of the eye.
  • These can also be used to cleanse wounds.
  • Finally, the sterile moist wipe dispenser includes 50 individually wrapped wipes which are suitable for cleaning wounds and the surrounding skin before applying a dressing.
  • The station also includes a plastic A5 frame and is supplied with 4 first aider inserts, allowing you to write the names and locations of up to five first aiders, so people know where to turn following an accident or illness.
  • Following personnel changes, these can easily be removed and replaced with a fresh insert.
  • Each component fits onto a well-organised, eye-catching poster with designated areas for each one alongside clear step-by-step first aid guidance with illustrations, giving easy access to lifesaving information including how to do the primary survey and perform CPR.
  • Wall fixings not supplied.
  • The Evolution First Aid Station, Large Contains:
    • First Aid Kit Contents:
      • 1x First Aid Guidance Leaflet
      • 6x HypaCover First Aid Dressings, 12 x 12 cm
      • 4x HypaCover First Aid Dressings, 18 x 18 cm
      • 4x HypaBand Triangular Bandages
      • 4x HypaCover Eye Dressings
      • 100x HypaPlast Washproof Plasters
      • 40x HypaClean Sterile Wipes
      • 3x HypaPlast Microporous Tape, 2.5 cm
      • 12x HypaTouch Nitrile Gloves (Pair)
      • 4x HypaCover Finger Dressings
      • 2x HypaGuard Face Shields
      • 3x HypaGuard Foil Blankets
      • 2x HypaSoothe Burn Dressings, 10 x 10 cm
      • 1x Clothing Cutters
      • 2x HypaBand Conforming Bandages, 7.5 cm
    • First Aid Station Contents:
      • 1x First Aid Kit
      • 2x First Aid Inspection Tags Tamper Proof Studs
      • 1x Salvequick Plasters Dispenser (40 Fabric Plasters 45 Washproof Plasters)
      • 1x HypaClens Eye Wash Dispenser (10 Eye Wash Pods, 20 mL)
      • 4x First Aider Location Sign (updateable inserts)
Specifications
Product Type First Aid Station
Material Plastic
Sterility Sterile