Learn More
Invitrogen™ Evolocumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559307
 View more versions of this product
Product Code. 30282796
  • £397.00 / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Evolocumab is a human IgG2 monoclonal antibody that binds to human PCSK9 (Proprotein Convertase Subtilisin Kexin Type 9).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Evolocumab
Recombinant Monoclonal
Unconjugated
AMG 145; Repatha
Protein A
RUO
Human
Antibody
IgG2SA
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized