Learn More
Thermo Scientific™ eVol™ XR Sample Dispensing System Kit
Product Code. 11826224 Shop All Thermo Scientific Products
Change view
Click to view available options
eVol digitally controlled analytical syringe
Product Code.
11826224 eVol digitally controlled analytical syringe
1 options
Product Code. 11826224 Supplier Thermo Scientific™ Supplier No. 66002024
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Get up and running quickly. The Thermo Scientific™ eVol™ Sample Dispensing System Kit includes an eVol XR dispensing system and three syringes (5, 50, and 500μL).
Get up and running quickly. The Thermo Scientific™ eVol™ Sample Dispensing System Kit includes an eVol XR dispensing system and three syringes (5, 50, and 500μL). The eVol Dispensing System is a unique, hand-held digital dispenser that accurately and reproducibly performs a wide variety of liquid handling procedures. The easy-to-use device couples an analytical syringe to a programmable electronic controller, enabling highly reproducible dispensing for all users.
The eVol Dispensing System is ideal for laboratories that need to repeatedly dispense nonaqueous liquids, offering greater accuracy and reproducibility for applications such as:
  • Routine dispensing of volatile solvents, hazardous, corrosive or viscous chemicals
  • Preparation of calibration standards
  • Addition of internal standards
The system unifies two precision devices: a digitally controlled electronic drive, and a Thermo Scientific™ XCHANGE™ enabled analytical syringe.
Repeatable Performance for Higher Confidence in Your Results
  • Can be calibrated easily by a user to ensure validity of results
  • Intuitive, easy-to-use interface with touch wheel and full-color screen
  • Complies with GLP and GMP guidelines
Exceptional Versatility and Functionality
  • Integrated XCHANGE coupling allows syringes to be quickly and easily changed, preventing possible cross-contamination
  • Addition of Thermo Scientific™ MEPS™ (Micro Extraction by Packed Sorbent) technology to an XCHANGE syringe enables automated sample preparation by SPE in a hand-held system
TRUSTED_SUSTAINABILITY
Specifications
Description Sample Dispensing System Kit
For Use With (Application) Routine Dispensing of Volatile Solvents, Hazardous, Corrosive or Viscous Chemicals
Product Type DispensingSystem
Product Line HyperSep
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.