Learn More
Thermo Scientific™ eVol™ XR Dispensing System Plungers
Product Code. 15260369 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
eVol System, 1000 μL
eVol System, 500 μL
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
15260369 eVol System, 1000 μL Each
13407036 eVol System, 500 μL Each
2 options
Product Code. 15260369
Brand: Thermo Scientific™ 66002041
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Order these plungers for use with the respective size syringe on the Thermo Scientific™ eVol­™ Dispensing System.
Use these Thermo Scientific™ eVol™ Dispensing System Plungers with the respective size syringe on the eVol Dispensing System. Like the syringes, these plungers are available in 5 to 1000μL sizes. The unique, hand-held eVol Dispensing System accurately and reproducibly performs a wide variety of liquid handling procedures. The easy-to-use device couples an analytical syringe to a programmable electronic controller, enabling highly reproducible dispensing for all users.
The eVol Dispensing System is ideal for laboratories that need to repeatedly dispense nonaqueous liquids, offering greater accuracy and reproducibility for applications such as:
  • Routine dispensing of volatile solvents, hazardous, corrosive or viscous chemicals
  • Preparation of calibration standards
  • Addition of internal standards
The system unifies two precision devices: a digitally controlled electronic drive, and a Thermo Scientific™ XCHANGE™ enabled analytical syringe.
Repeatable Performance for Higher Confidence in Your Results
  • Can be calibrated easily by a user to ensure validity of results
  • Intuitive, easy-to-use interface with touch wheel and full-color screen
  • Complies with GLP and GMP guidelines
Exceptional Versatility and Functionality
  • Integrated XCHANGE coupling allows syringes to be quickly and easily changed, preventing possible cross-contamination
  • Addition of Thermo Scientific™ MEPS™ (Micro Extraction by Packed Sorbent) technology to an XCHANGE syringe enables automated sample preparation by SPE in a hand-held system
TRUSTED_SUSTAINABILITY
Specifications
Product Type Dispensing System Plunger
Product Line HyperSep
For Use With (Equipment) eVol System, 1000 μL
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.