missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Evodiamine

Product Code. 16471486
Encompass_Preferred
Change view
Click to view available options
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16471486 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16471486 Supplier MedChemExpress Supplier No. HYN0114

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Evodiamine is an alkaloid isolated from the fruit of Evodia rutaecarpa Bentham with diverse biological activities including anti-inflammatory, anti-obesity, and antitumor.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 303.36 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 518-17-2
Product Type Natural Products
Purity 99.59%
Solubility DMSO : 10 mg/mL (32.96 mM∣Need ultrasonic)
Synonym (+)-Evodiamine∣ d-Evodiamine
Quantity 200 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.