missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EVL Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569099
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.
Ena/VASP proteins are actin-associated proteins involved in a range of processes dependent on cytoskeleton remodeling and cell polarity such as axon guidance and lamellipodial and filopodial dynamics in migrating cells. EVL enhances actin nucleation and polymerization.
Specifications
| EVL | |
| Polyclonal | |
| Unconjugated | |
| EVL | |
| AI528774; ena/vasodilator-stimulated phosphoprotein-like; Ena/VASP-like protein; Enah/Vasp-like; Ena-vasodilator stimulated phosphoprotein; Evl; Rnb6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14026 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P70429 | |
| EVL | |
| synthetic peptide directed towards the middle region of mouse EVL. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction