missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EVL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118886
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total EVL.
Ena/VASP proteins are actin-associated proteins involved in a range of processes dependent on cytoskeleton remodeling and cell polarity such as axon guidance and lamellipodial and filopodial dynamics in migrating cells. EVL enhances actin nucleation and polymerization.
Specifications
| EVL | |
| Polyclonal | |
| Unconjugated | |
| EVL | |
| AI528774; ena/vasodilator-stimulated phosphoprotein-like; Ena/VASP-like protein; Enah/Vasp-like; Ena-vasodilator stimulated phosphoprotein; Evl; Rnb6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14026, 51466, 79115 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O08719, P70429, Q9UI08 | |
| EVL | |
| A synthesized peptide derived from human EVL(Accession Q9UI08), corresponding to amino acid residues E293-T343. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction