missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
EVI5L Polyclonal specifically detects EVI5L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | EVI5L |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500-1:1000 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Alias | ecotropic viral integration site 5-like, Ecotropic viral integration site 5-like protein, EVI5-like protein |
| Gene Symbols | EVI5L |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:VVRQQCSSAAEDLQKAQSTIRQLQEQQENPRLTEDFVSHLETELEQSRLRETETL |
| Show More |
For Research Use Only
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?