missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EVI5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-40620
This item is not returnable.
View return policy
Description
EVI5 Polyclonal specifically detects EVI5 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).
Specifications
| EVI5 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| dJ846D11.1 (ecotropic viral integration site 5), ecotropic viral integration site 5, ecotropic viral integration site 5 protein homolog, EVI-5, NB4SNeuroblastoma stage 4S gene protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 7813 | |
| Store at 4C. Do not freeze. |
| Immunoprecipitation, Western Blot | |
| 1.0 mg/mL | |
| Immunoprecipitation 2-5 ug/mg lysate, Western Blot (Negative) | |
| NP_005656.4 | |
| EVI5 | |
| The immunogen recognized by this antibody maps to a region between residue 525 and 575 of human Ecotropic Viral Integrations Site 5 using the numbering given in entry NP_005656.4 (GeneID 7813). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction