missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EVI5 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-40620AF594
This item is not returnable.
View return policy
Description
EVI5 Polyclonal antibody specifically detects EVI5 in Human samples. It is validated for Western Blot
Specifications
| EVI5 | |
| Polyclonal | |
| Western Blot | |
| dJ846D11.1 (ecotropic viral integration site 5), ecotropic viral integration site 5, ecotropic viral integration site 5 protein homolog, EVI-5, NB4SNeuroblastoma stage 4S gene protein | |
| The immunogen recognized by this antibody maps to a region between residue 525 and 575 of human Ecotropic Viral Integrations Site 5 using the numbering given in entry NP_005656.4 (GeneID 7813). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 7813 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction