missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EVI2B Antibody (MEM-216), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62207MFV450
This item is not returnable.
View return policy
Description
EVI2B Monoclonal antibody specifically detects EVI2B in Human samples. It is validated for Flow Cytometry
Specifications
| EVI2B | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 2124 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| MEM-216 | |
| Flow Cytometry | |
| CD361 antigen, D17S376, ecotropic viral integration site 2B, Ecotropic viral integration site 2B protein homolog, EVDBprotein EVI2B, EVI-2B | |
| Raji cells | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction