missing translation for 'onlineSavingsMsg'
Learn More

EVI2B Antibody, DyLight 405, Novus Biologicals™

Product Code. 18319192 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18319192 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18319192 Supplier Novus Biologicals Supplier No. NBP299056V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EVI2B Polyclonal specifically detects EVI2B in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EVI2B
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias CD361 antigen, D17S376, ecotropic viral integration site 2B, Ecotropic viral integration site 2B protein homolog, EVDBprotein EVI2B, EVI-2B
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of mouse EVI2B.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2124
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.