missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
EVI2B Polyclonal specifically detects EVI2B in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | EVI2B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 594 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD361 antigen, D17S376, ecotropic viral integration site 2B, Ecotropic viral integration site 2B protein homolog, EVDBprotein EVI2B, EVI-2B |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of mouse EVI2B. |
| Purification Method | Antigen Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?