Learn More
Invitrogen™ EVI1 Recombinant Rabbit Monoclonal Antibody (11H41L13)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ 702005
Description
This antibody is predicted to react with Monkey, Rabbit and Bat Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
EVI1 (MECOM) is a transcriptional regulator and oncoprotein that may be involved in hematopoiesis, apoptosis, development, cell differentiation, and proliferation. It can interact with CTBP1, SMAD3, CREBBP, KAT2B, MAPK8, and MAPK9. The EVI1 gene can undergo translocation with the AML1 gene, resulting in overexpression of the EVI1 gene and the onset of leukemia. Several transcript variants encoding a few different isoforms have been found for this gene.
Specifications
| EVI1 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q03112 | |
| MECOM | |
| Peptides corresponding to Human MECOM (aa 367-384, 901-918). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 11H41L13 | |
| Unconjugated | |
| MECOM | |
| AML1-EVI-1; AML1-EVI-1 fusion protein; D630039M04Rik; ecotropic viral integration site 1; ecotropic virus integration site 1 protein; Ecotropic virus integration site 1 protein homolog; EVI1; Evi-1; Histone-lysine N-methyltransferase MECOM; Jbo; Mds; MDS1; MDS1 and EVI1 complex locus; MDS1 and EVI1 complex locus protein; MDS1 and EVI1 complex locus protein EVI1; MDS1 and EVI1 complex locus protein MDS1; Mds1-Evi1; Mecom; MGC163392; MGC97004; myelodysplasia syndrome 1 homolog; Myelodysplasia syndrome 1 protein; Myelodysplasia syndrome 1 protein homolog; myelodysplasia syndrome-associated protein 1; oncogene EVI1; OTTHUMP00000213163; OTTHUMP00000213164; OTTHUMP00000213165; OTTHUMP00000213187; OTTHUMP00000213192; PR domain 3; PRDM3; RUSAT2; zinc finger protein Evi1; Znfpr1b1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2122 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.