missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EVI1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23201-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a transcriptional regulator and oncoprotein that may be involved in hematopoiesis, apoptosis, development, and cell differentiation and proliferation. The encoded protein can interact with CTBP1, SMAD3, CREBBP, KAT2B, MAPK8, and MAPK9. This gene can undergo translocation with the AML1 gene, resulting in overexpression of this gene and the onset of leukemia. Several transcript variants encoding a few different isoforms have been found for this gene.Specifications
| EVI1 | |
| Polyclonal | |
| Unconjugated | |
| MECOM | |
| AML1 EVI 1, EVI 1, EVI1, MDS1 EVI1, MECOM, PRDM3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2122 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q03112 | |
| MECOM | |
| EVI1 Fusion Protein Ag19603 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction