missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EVI-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48848
This item is not returnable.
View return policy
Description
EVI-1 Polyclonal antibody specifically detects EVI-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| EVI-1 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| AML1-EVI-1, AML1-EVI-1 fusion protein, ecotropic viral integration site 1, Ecotropic virus integration site 1 protein homolog, EVI-1, EVI1MDS1-EVI1, MDS1, MDS1 and EVI1 complex locus, MDS1 and EVI1 complex locus protein EVI1, MDS1 and EVI1 complex locus protein MDS1, MGC163392, MGC97004, myelodysplasia syndrome 1, Myelodysplasia syndrome 1 protein, Myelodysplasia syndrome-associated protein 1, oncogene EVI1, PRDM3, zinc finger protein Evi1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: ATSSPHSELESTGAILDDKEDAYFTEIRNFIGNSNHGSQSPRNVEERMNGSHFKDEKALVTSQNSDLLDDEEVEDEVLLDEEDEDNDITGKTGKEPVTSNLHEGNPEDDYEETSALEMSCKTSPVRYKEEEY | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 2122 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction