missing translation for 'onlineSavingsMsg'
Learn More

Everolimus

Product Code. 15492959
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492959 25 mg 25mg
15482959 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15492959 Supplier Thermo Scientific Alfa Aesar Supplier No. J60139.MB
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A rapamycin analog | CAS: 159351-69-6 | C53H83NO14 | 958.24 g/mol

Everolimus is a rapamycin analog. It is used to treat certain types of kidney cancer, breast cancer and brain tumor. It prevents transplant rejection by decreasing the activity of the immune system. It also act as a kinase inhibitor. It is used in the treatment of subependymal giant cell astrocytoma (SEGA; a type of brain tumor) in adults and children 1 year of age and older who have TSC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dimethysulfoxide,ethanol and chloroform. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 159351-69-6
Molecular Formula C53H83NO14
Molecular Weight (g/mol) 958.24
MDL Number MFCD07785165
InChI Key HKVAMNSJSFKALM-PWMUVMERSA-N
PubChem CID 129317877
SMILES CC1CCC2CC(C(=CC=CC=CC(CC(C(=O)C(C(C(=CC(C(=O)CC(OC(=O)C3CCCCN3C(=O)C(=O)C1(O2)O)C(C)CC4CCC(C(C4)OC)OCCO)C)C)O)OC)C)C)C)OC

Specifications

Color White
Quantity 25 mg
Merck Index 14,3907
Solubility Information Soluble in dimethysulfoxide,ethanol and chloroform; Slightly soluble in water.
Formula Weight 958.22
Physical Form Powder
Chemical Name or Material Everolimus

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.