missing translation for 'onlineSavingsMsg'
Learn More

A&D Weighing EK-i Series Compact Balance
GREENER_CHOICE

Product Code. 16601022
Change view
Click to view available options
Capacity:
12,000 g
1200 g
200 g
2000 g
300 g
3000 g
400 g
4000 g
6000 g
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
16601022 4000 g Each
16611012 12,000 g Each
16611022 400 g Each
16621022 6000 g Each
16631012 2000 g Each
16641012 200 g Each
16661012 3000 g Each
16681012 300 g Each
16691002 1200 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 16601022 Supplier A&D Weighing Supplier No. EK4100i
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

EK-i Series Compact Balances feature a compact rugged design, reliable SG weighing technology, and versatile single and triple weighing ranges.

  • EC Type approved (EK-610i-EC, EK-6100i-EC)
  • Standard RS-232C interface
  • Large LCD display with backlight
  • Compact design
  • GLP compliant
  • Overload protection
  • Standard comparator function
  • Optional rechargeable battery pack
  • 9 Weighing units plus count and percentage mode
  • Counting function with Automatic Counting Accuracy Improvement (ACAI)
  • Full digital calibration with optional user definable mass setting

Specifications

Certifications/Compliance GLP/GMP/ISO
Connection Type RS-232 C
Capacity 4000 g
Display Type Backlit LCD
Readability 0.1 g
Material ABS, Stainless Steel
Model EK-4100i
Electrical Requirements 110/240 V to adapter (9 V, 670 mA), Ni-MH battery Pk. (option)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.