missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EVER2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen OST00157W
Code : LD
Additional Details : Weight : 0.99000kg
Description
Glycerol (1:1) may be added for additional stability. Reconstitute in 100 μL of sterile water.
Specifications
EVER2 | |
Polyclonal | |
Tmc8 | |
TMC8, Epidermodysplasia verruciformis protein 2, EVER2, EVIN2 | |
Rabbit | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q7TN58 | |
TMC8 | |
A synthetic peptide from the 1t cytoplasmic loop of mouse transmembrane channel-like protein 8 | |
RUO | |
217356 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |