missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eve Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569510
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Fruit fly: 100%.
eve gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
Specifications
| eve | |
| Polyclonal | |
| Unconjugated | |
| eve | |
| 10.5; 10.9; 14.10; 20.35; CG2328; CG2328-PA; complementation group F; Dmel\CG2328; Dmel_CG2328; dm-eve; E(eve); Eve; eve PRE/TRE element; eve2; even; even skipped; evenskip; even-skiped; evenskipped; even-skipped; eve-PA; F; group V; group VI; l(2)46Ce; l(2)46CFg; l(2)46CFh; l(2)46CFj; l(2)46CFp; l(2)46Cg; lethal(2)46Ce; Segmentation protein even-skipped; V; VI | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 36039 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P06602 | |
| eve | |
| synthetic peptide corresponding to a region of fruit fly eve (aa 1-50). | |
| 100 μL | |
| Primary | |
| Drosophila | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction