missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EVC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31268
This item is not returnable.
View return policy
Description
EVC2 Polyclonal specifically detects EVC2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| EVC2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| EVC2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human EVC2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 132884 | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| Ellis van Creveld syndrome 2, LBNlimbin | |
| Rabbit | |
| 148 kDa | |
| 100 μL | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Mouse (85%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction