missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EVC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571411
This item is not returnable.
View return policy
Description
EVC Polyclonal Antibody for Western Blot
This gene encodes a protein containing a leucine zipper and a transmembrane domain. This gene has been implicated in both Ellis-van Creveld syndrome (EvC) and Weyers acrodental dysostosis.
Specifications
| EVC | |
| Polyclonal | |
| Unconjugated | |
| Evc | |
| DWF-1; Ellis van Creveld gene homolog; Ellis van Creveld gene syndrome; Ellis van Creveld protein; Ellis van Creveld syndrome; ellis-van Creveld syndrome protein; Ellis-van Creveld syndrome protein homolog; EVC; EvC ciliary complex subunit 1; EvC protein (Ellis-van Creveld syndrome); EVC1; EVCL | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2121 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P57679 | |
| Evc | |
| synthetic peptide directed towards the N terminal region of human EVC. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction