missing translation for 'onlineSavingsMsg'
Learn More

Evacetrapib, MedChemExpress

Product Code. 16465648
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16465648 10 mg 10mg
16456166 100 mg 100mg
16451817 50 mg 50mg
16418347 5 mg 5mg
4 options
This item is not returnable. View return policy

Product Code. 16465648

Brand: Medchem Express HY13327

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Evacetrapib is a potent and selective of CETP inhibitor, which inhibits human recombinant CETP protein (IC50 5.5 nM) and CETP activity in human plasma (IC50 36 nM) in vitro.

Evacetrapib is a potent and selective of CETP inhibitor, which inhibits human recombinant CETP protein (IC50 5.5 nM) and CETP activity in human plasma (IC50 36 nM) in vitro.

Specifications

Chemical Name or Material Evacetrapib
Physical Form Solid
CAS 1186486-62-3
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C31H36F6N6O2
For Use With (Application) COVID-19-immunoregulation
Synonym LY2484595
Solubility Information DMSO : ≥ 100 mg/mL (156.58 mM)
SMILES O=C([C@H]1CC[C@H](CN2CCC[C@H](N(CC3=CC(C(F)(F)F)=CC(C(F)(F)F)=C3)C4=NN(C)N=N4)C5=CC(C)=CC(C)=C52)CC1)O
Molecular Weight (g/mol) 638.65
Formula Weight 638.65
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.