missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EVA1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569180
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Dog: 89%; Goat: 80%; Guinea Pig: 87%; Horse: 100%; Human: 100%; Mouse: 82%; Rabbit: 86%; Rat: 93%; Yeast: 100%.
EVA1B is a protein coding gene.
Specifications
| EVA1B | |
| Polyclonal | |
| Unconjugated | |
| EVA1B | |
| 2610027C15Rik; C1orf78; eva-1 homolog B; eva-1 homolog B (C. elegans); EVA1B; Fam176b; family with sequence similarity 176, member B; protein eva-1 homolog B; Protein FAM176B; RGD1308876 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 362597 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| EVA1B | |
| synthetic peptide directed towards the middle region of human EVA1B. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction