missing translation for 'onlineSavingsMsg'
Learn More

EVA1/MPZL2 Antibody (2E10), Novus Biologicals™

Product Code. 18319709 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18319709 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18319709 Supplier Novus Biologicals Supplier No. H00010205M06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

EVA1/MPZL2 Monoclonal antibody specifically detects EVA1/MPZL2 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen EVA1/MPZL2
Applications ELISA, Immunocytochemistry, Sandwich ELISA
Classification Monoclonal
Clone 2E10
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1:10-1:500, Sandwich ELISA 1:100-1:2000
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005788
Gene Alias EVAEpithelial V-like antigen 1EVA1myelin protein zero-like protein 2, myelin protein zero-like 2
Host Species Mouse
Immunogen MPZL2 (NP_005788.1, 27 a.a. ~ 136 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VEIYTSRVLEAVNGTDARLKCTFSSFAPVGDALTVTWNFRPLDGGPEQFVFYYHIDPFQPMSGRFKDRVSWDGNPERYDASILLWKLQFDDNGTYTCQVKNPPDVDGVIG
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10205
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.