missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EV71 VP1, Mouse anti-Viruses, Clone: 4D3, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB1255-M08.100ug
This item is not returnable.
View return policy
Description
Sequence: GDRVADVIESSIGDSVSRALTQALPAPTGQNTQVSSHRLDTGEVPALQAAEIGASSNTSDESMIETRCVLNSHSTAETTLDSFFSRAGLVGEIDLPLEGTTNPNGYANWDIDITGYAQMRRKVELFTYMRFDAEFTFVACTPTGGVVPQLLQYMFVPPGAPKPDSRESLAWQTATNPSVFVKLTDPPAQVSVPFMSPASAYQWFYDGYPTFGEHKQEKDLEYGACPNNMMGTFSVRTVGSSKSKYPLVVRIYMRMKHVRAWIPRPMRNQNYLFKANPNYAGNSIKPTGTSRTAITTLSpecifications
| EV71 VP1 | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.4 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| ELISA, Western Blot | |
| 4D3 | |
| The optimal working dilution should be determined by the end user. | |
| AY258310 | |
| EV71 VP1 (AAP82198.1, 1 a.a. ∼ 297 a.a) recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 κ |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- EV71 VP1 monoclonal antibody, clone 4D3
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P103-Read label before use.
- P233-Keep container tightly closed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P404-Store in a closed container.
Supplemental information
- MIXTURE LIST-Contains : sodium phosphate dibasic, potassium phosphate monobasic
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction