missing translation for 'onlineSavingsMsg'
Learn More

TDS 442 Standard Solution, Eutech™

Product Code. 11549124
Click to view available options
Chemical Name or Material:
442 Calibration Solution
442 Standard Solution
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 11549124

Brand: Thermo Scientific Eutech 01X109105

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For TDS calibration and standards, choose from our available Eutech™'s TDS Standards, Calibrations, and Solutions

Eutech™ TDS Standard Solutions are used as standards for TDS meters. Get quick and accurate conductivity TDS measurements using Eutech™ TDS meters in the field, laboratories, and schools.

Features:

  • Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 442 Calibration Solution
Packaging Plastic Bottle
Physical Form Liquid
Color Colorless
pH 7
Boiling Point ∼100°C
Solution Type Standard Solution
Type TDS Standard
Industry Type Laboratory Use
Quantity 480 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.