Learn More
Thermo Scientific™ Eutech™ Salt Bridge for pH/ORP Differential Electrodes ECFP21A1A1 and ECFR21A1A1
Description
Eutech™ Salt Bridge is used for pH/ORP Differential Electrodes ECFP21A1A1 and ECFR21A1A1. The electrodes are designed for continuous use in industrial process applications.
Features:
- A replaceable heavy-duty quad junction salt bridge for extended life in severe applications.
Specifications
Specifications
| For Use With (Equipment) | pH/ORP Differential Electrodes ECFP21A1A1, ECFR21A1A1 |
| Type | Salt Bridge |