missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Eutech™ pHTestr 10BNC pH Meter
Description
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. Eutech™ pH Testr 10 BNC pocket testers are ideal choices for quick and accurate pH checks and routine applications in laboratories, productions plants, and schools.
Features:
- Large Display Screen - dual display LCD for enhanced readability, provides comprehensive meter information
- Waterproof, Dustproof Housing - IP67 rated, maintains the integrity of your tester even in harsh conditions. Plus it floats!
- User-Selectable Electrodes with BNC connectors - for greater flexibility with choice of electrodes for specific applications
- Push-Button Calibration with Auto-Buffer Recognition - quick, easy calibration with no calibration mistakes
- Choice of USA or NIST pH buffer standards with up to 3 points
- Auto-Power Off saves battery power after non-use
- Hold Function freezes reading momentarily for easy viewing
- Self-Diagnostic with message codes for easy troubleshooting
Applications
- General: Ideal for quick and accurate pH checks in pools and spas, aquariums and hydroponics operations, or anywhere frequent pH testing is required.
- Industrial: Use for cooling towers, food processing water testing and process/wastewater testing in metal finishing, photo development, printing and chemical industries.
- Educational: Useful in schools, many laboratory applications and ecology studies.
Specifications
Specifications
| Description | Waterproof pHTestr 10 BNC with MTC; BNC connector sensor; ± 0.1 pH resolution (meter only – electrode to be ordered separately) |
| Includes | Large Dual Custom Display, Self-diagnostic; Hold function; Auto-Power Off after 8.5 minutes |
| Accuracy (pH) | ±0.01 |
| Calibration | Automatic |
| Range (pH) | -1 to 15 |
| Resolution (pH) | 0.01 |
| Calibration Points | 3 |
| Display Type | LCD, Dual Display with Battery Indicator, Error Messages/Codes |
| Accuracy | ±0.1 pH |
| Model | pHTestr 10BNC |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?