Learn More
Thermo Scientific™ Eutech™ pHSpear pH Meter
Description
Eutech™ pHSpear pH Meter is a unique pHTestr with a spear electrode. The Eutech waterproof pH Spear with tough, spear tip open pore sensor is a rugged, user-friendly tester ideal for direct measurement of solid or semi-solid samples such as cheese, meats, fruits, bread and other similar samples. More importantly, the pH Spear is an easy-to-clean, fuss-free pH tester specially designed for the food professional. Unlike conventional pH electrodes, the durable glass tip electrode, enclosed in a resilient engineered plastic body is ideal for piercing solid or semi-solid food samples in direct measurement. Open pore reference junction minimises clogging and delivers fast, stable measurements. Double junction sensor prolongs useful life of electrode with no contamination of samples. Maintenance is easy. Simply wash with mild detergent.
Features
- Large Display Screen - 27mm (H) x 21mm(W) large LCD displaymakes it easy to read even if tester is held at a distance.
- One Hand Operation - One hand operation is possible with wireless electrode. Eyelet allows you to attach a lanyard for greater portability.
- Waterproof, Unique Ribbed Design - IP67 waterproof housing protects against high humidity or condensation. Unique ribbed design ensures firmer grip when wet or greasy.
- User-Replaceable Sensor - With the replaceable electrode, you can reuse the tester and savemoney over time. Twist-off sensor design allows easy replacement of electrode anytime.
Applications
pH is one of the key parameters in food production formeasurement of solid or semi-solid sample such as cheese,meats, fruits, poultry, bread or other similar samples.
Specifications
- pH Range: -1.00 to 15.00 pH
- Resolution: 0.01 pH
- Accuracy: ±0.01 pH
- No. of Calibration Points: Up to 3 points
- pH Buffer Options: USA - pH 4.01, 7.00, 10.01; NIST - pH 4.01, 6.86, 9.18
- LCD Display: Dual display with battery indicator, errormessages/codes
- Special Functions: Large Dual Custom Display, Self-diagnostic; Hold function; Auto-Power Off after 8.5minutes
- Power: 4 x 1.5V alkaline button cell batteries; greater than 500 hrs
Specifications
Specifications
| Description | Waterproof pHTestr with spear electrode |
| Junction | Double |
| Material | Plastic |
| Type | pH Meter |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.