missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ ECFG7370101B pH electrode

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. 

Brand:  Thermo Scientific™ 93X218819

218.37 GBP valid until 2024-04-30
Use promo code "21656" to get your promotional price.



Code : A0

Additional Details : Weight : 0.12600kg

Product Code. 11533885

  • £232.33 / Each
In Stock
to see stock.
Add to basket

Description

Description

The measure of a liquid acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications. Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. This clog free double-junction glass pH electrode is ideal for samples containing TRIS buffers, sulfides and more. Easy to operate fill hole sleeve mechanism is used for general pH measurements. Available with 1 m cable.

Specifications

  • Parameter: pH
  • Range: 0 to 13 pH
  • Temperature Range: 0 to 100 °C
  • Liquid Junction Type: Annular ceramic
  • Internal Reference Type: Ag/AgCl
  • Sealed/Refillable: Refillable
  • Reference Junction: Double
  • Refilling Reference Electrolyte: ECRE002
  • Dimensions (Shaft): 110 x 12mm
  • Cable Length: 1m
  • Connector: BNC
  • Used With: All pHmeters with BNC input connector
  • Specifications

    Specifications

    BNC
    0 to 13
    pH Electrode
    Double
    0°C to 100°C
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers