Learn More
Eutech™ pH Buffers and Solutions, Color-Coded
Description
The measure of a liquid's acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications.
Eutech™ offers a wide range of pH buffers and solutions for use with pH meters and electrodes.
Features:
- Buffers and calibration solutions packed in 60mL, 480 ml or 1L durable plastic bottles – economical and convenient for laboratory use.
Buffer Color Codes:
- Pink = pH 4.01
- Yellow = pH 7.00
- Blue = pH 10.01
Specifications
Specifications
| Color | Red, Yellow, Blue |
| Description | pH Buffer Kit |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 4.01, 7.00, 10.01 |
| Quantity | 480 mL Ea. |
| Solution Type | pH Buffer |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.