missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eutech pH Buffer Sachets
Click to view available options
Chemical Name or Material:
pH Buffer Sachet
pH Buffer Sachet Set, Deionized Water
pH:
4.01
4.01, 7.00, 10.01
7
Unit Size:
Each
Pack of 20
Description
pH Buffer Sachets are NIST-traceable sachets for quick, accurate and convenient calibrations. Simply insert electrode into sachet pack, calibrate, rinse and discard sachet – no separate container needed.
Features:
- pH Buffer Sachets -20 ml
- NIST Traceable
Specifications
Specifications
| Chemical Name or Material | pH Buffer Sachet |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 4.01 |
| Quantity | 20 x 20 mL |
| Solution Type | Buffer |
| Certifications/Compliance | NIST Traceable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction