missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ pH 5+ meter

Product Code. p-7114752
Click to view available options
Type:
pH Meter with 3-in-1 Electrode
pH Meter with ATC Electrode
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11529164

Brand: Thermo Scientific™ 01X244911

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.

The measure of a liquid's acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications. Eutech™ offers a wide range of pH meters and electrodes and Eutech™ pH 5+ Meter is an ideal choice for pH applications.

Features:

  • Multi-Point Calibration (up to 5 points) with Auto-Buffer Recognition - choice of USA, NIST or pure water buffer standards with ±0.01 pH accuracy
  • Automatic Temperature Compensation (ATC) for the highest accuracy under fluctuating temperatures
  • Hold Function momentarily freezes reading for easy viewing and recording
  • Auto-Power Off saves battery power after non-use
  • Self-Diagnostic with message codes for easy troubleshooting
  • Large Custom LCD provides optimum viewing even at a distance
  • Protective Rubber Boot shields meter from accidental knocks and features a sturdy built-in stand for easy bench-top operation
  • Rugged Carrying Case (Optional) provides handy on-site meter calibration and measurement
  • pH Buffer Options: 1.68, 4.01, 7.00, 10.01, 12.45 (USA), 1.68, 4.01, 6.86, 9.18, 12.45 (NIST), 4.10, 6.97 (Pb)
TRUSTED_SUSTAINABILITY

Specifications

Description Eutech pH 5+ Meter with ATC Electrode ECPHTEM01P, Rubber Armour/Stand
Includes Auto Power-Off after 17 minutes; Hold & Self-diagnostic messages
Accuracy (Temperature) ±0.5°C
Accuracy (pH) ±0.01
Range (pH) 0 to 14
Resolution (pH) 0.01
Calibration Points 2 to 5
Temperature Range (Metric) 0°C to 100°C
Resolution (Temperature) 0.1°C
Display Type LCD
Temperature Compensation Automatic/Manual (0°C to 100°C)
Battery Type 4 AAA x 1.4V batteries
Type pH Meter with ATC Electrode
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.