missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ pH 450 Meter

Product Code. 15327053
Change view
Click to view available options
Type:
pH Meter
pH Meter and Electrode
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15327053 pH Meter and Electrode Each
15317053 pH Meter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15327053 Supplier Thermo Scientific™ Supplier No. 01X651629
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.

Eutech™ offers a wide range of pH meters and electrodes. pH 450 Meter Kit is a rugged waterproof meter ideal for any environment and offers measurement of pH / mV / Ion / Temperature.

Features

  • Durable waterproof and dustproof design with IP67 rating
  • Large, backlit display – View measurement with temperature and % slope
  • Grip-Clip™ – Quickly and easily secures the pH electrode to your beaker or container
  • Calibrate up to 5 points automatically with USA or NIST buffer groups (up to 6 points with DIN). New manual option allows calibration with up to 5 custom buffer standards
  • Auto buffer recognition – Automatically identifies the correct pH buffer for rapid calibration
  • Calibration due alarm – Set a visible reminder when it is time to recalibrate the meter
  • Ready indicator, manual or automatic measurement hold feature and optional automatic shutoff function
  • Selectable ºC/ºF – Choose units to best fit your application
  • Intuitive, user-friendly icons – Electrode status displays pH slope
  • Store up to 500 data sets – Easy USB or RS232 output with optional data cables
  • Built-in real-time clock stamps stored data and calibration data with time and date – Meets standards for GLP (Good Laboratory Practices)
  • Extended battery life from two AA batteries, or use optional universal power adapter
  • Built-in stand with wall-mount option – Allows for easy benchtop measurement or storage
  • Compatible with pH, ORP or ISE electrodes having a BNC connector
TRUSTED_SUSTAINABILITY

Specifications

Description pH 450 Meter & Electrode: Meter with double junction pH electrode ECFC7252201B and ATC probe PHWPTEM01W, Grip-Clip™, two AA batteries
Includes Real-Time Clock: Time and date stamp on calibration and stored data
Accuracy (Temperature) ±0.5°C/±0.9°F
Accuracy (mV) ±0.2 +2 LSD or +0.5 % of reading, whichever is greater
Accuracy (pH) ±0.01
Outputs USB, RS-232
Measurement Modes 500
Calibration mV: ±200 mV, Ion: From 2 to 4 or 2 to 5 points: 0.10, 1.00, 10.0, 100.0 & 1000 / 0.20, 2.00, 20.0
Range (pH) -2 to 16
Resolution (pH) 0.01
Range (mV) ±2000
Resolution (mV) 0.1 mV ±999.9 mV; 1 mV beyond
Calibration Points Up to 5 (USA & NIST & CUSTOM), Up to 6 (DIN)
Temperature Range (Metric) -10°C to +110°C
Temperature Range (English) 14.0°F to 230.0°F
Resolution (Temperature) 0.1°C/°F
Temperature Probe Calibration Offset in 0.1 increments, ±10 °C or ±18 °F
Display Type LCD, Custom with Backlight
Memory Stores up to 500 data sets
Data Points 500
IP Rating IP67
AC Adapter Optional
Dimensions (L x W x H) Meter: 7 7/8 x 3 1/8 x 2 1/4 in (20 x 8.3 x 5.7 cm), Boxed: 9 1/8 x 9 1/8 x 2 3/4 in (23 x 23 x 7 cm)
Weight (Metric) Meter: 0.5 kg, Boxed: 0.7 kg
Weight (English) Meter: 1 lb., Boxed: 1.4 lb.
Range (Ion) 0.0 to 2000 ppm
Resolution (Ion) 0.01 / 0.1 / 1 ppm
Temperature Compensation Automatic/Manual (-10°C to 110°C)
Battery Type 2 x AA batteries (included)
Type pH Meter and Electrode
Battery Life >500 hr. (without backlight)
Accuracy (Ion) ±0.5 % full scale (monovalent); ±1 % full scale (divalent)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.