missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Eutech™ CyberScan pH 610 Meter
Click to view available options
Range (pH):
-2 to 20
Unit Size:
Each
Description
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. Eutech™ CyberScan pH 610 Meter is an ideal choice for routine applications in laboratories, productions plants and schools.
Features:
- 6 calibrations points and 20 auto/manual buffer options sets for quick, convenient calibration.
- Higher accuracy under varying temperature with Automatic/ Manual Temperature Compensation.
- Mega-memory space that stores/ recalls up to 500 Data Sets with temperature readings, with time and date-stamping formatted to meet GLP standards.
- Wireless and cable-free data transfer with infrared IrDA technology. Comes with PC-based Complimentary Data Acquisition Software for hassle-free data import and report-generation into any popular spreadsheet programs of your choice.
- Recall Electrode Slope, Electrode Offset Value and Previous Calibration Values for easy reference
- Large, Easy-to-Read Custom Display with simultaneous readings provides optimum viewing with user-friendly icons
- Hold Function momentarily freezes reading for easy viewing and recording
- Self-Diagnostic with message codes for easy troubleshooting
- Volatile Memory holds settings and stored data even when you run out of batteries.
- pH Buffer Options: USA, NIST, DIN, PWB
Specifications
Specifications
| Description | pH610 pH/mV Meter with ATC Probe & DAS software |
| Includes | Stability Indicator; Self-diagnostic; Non-Volatile Memory; Hold & Auto-Power Off after 20 minutes |
| Accuracy (Temperature) | 0.1°C/1.0°F |
| Accuracy (mV) | ±0.2 |
| Accuracy (pH) | ±0.002 |
| Outputs | IrDA, RS-232C |
| Measurement Modes | 500 |
| Calibration | Custom Buffer Calibration |
| Range (pH) | -2 to 20 |
| Resolution (pH) | 0.001, 0.01, 0.1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction