missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech Handheld Meters Calibration Solution, 1413μS/cm KCl

Product Code. 12385168
This item is not returnable. View return policy

Product Code. 12385168

Brand: Thermo Scientific™ 01X211207

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Periodic calibration with accurate standards is recommended for meters

  • 1413μS/cm KCl calibration solution
  • Solutions packed in 480mL durable plastic bottles
  • Economical and convenient for laboratory use.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Inorganic Salts
Name Note Nil
Packaging 1 case
CAS 7447-40-7
Specific Gravity 1
Physical Form Liquid
Molecular Formula Complex
Concentration 1413 μS/cm calibration solution
Color Colorless
pH 7.0
Boiling Point 102.4°C
For Use With (Equipment) Eutech Handheld Meters
Solution Type Calibration Solution
Type Calibration Solution
Quantity 450 mL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.