missing translation for 'onlineSavingsMsg'
Learn More

Eurycomanone, MedChemExpress

Product Code. 16461748
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16461748 5 mg 5mg
16452599 1 mg 1mg
16449308 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16461748 Supplier Medchem Express Supplier No. HYN5012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Eurycomanone could increases spermatogenesis by inhibiting the activity of phosphodiesterase and aromatase in steroidogenesis.

Eurycomanone could increases spermatogenesis by inhibiting the activity of phosphodiesterase and aromatase in steroidogenesis.

Specifications

Chemical Name or Material Eurycomanone
CAS 84633-29-4
Color White
Purity Grade Notes Research
Molecular Formula C20H24O9
Quantity 5 mg
Synonym Pasakbumin A
Solubility Information DMSO : 16.67 mg/mL (40.82 mM; Need ultrasonic)
SMILES O[C@@]1([C@H]2O)[C@@]34[C@@]([C@](OC4)(O)[C@H](O)C1=C)([H])[C@]([C@@](C(C)=CC5=O)([H])C[C@@]3([H])OC2=O)([C@@H]5O)C
Molecular Weight (g/mol) 408.4
Formula Weight 408.4
Percent Purity 98.02%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.