missing translation for 'onlineSavingsMsg'
Learn More

Europium solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™

Product Code. 12927353 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Bottle
Unit Size:
100mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
12927353 50 mL Bottle 50mL
12937353 100 mL Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12927353 Supplier Fisher Chemical Supplier No. S 618 050

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

CAS 7440-53-1
Molecular Formula Eu
Molecular Weight (g/mol) 151.96
MDL Number MFCD00010992
InChI Key OGPBJKLSAFTDLK-UHFFFAOYSA-N
PubChem CID 23981
ChEBI CHEBI:32999
IUPAC Name europium
SMILES [Eu]

Specifications

Packaging Bottle
Concentration 10 mg/L
Solvent or Matrix Nitric Acid
Quantity 50 mL
Chemical Name or Material Europium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.