missing translation for 'onlineSavingsMsg'
Learn More

Europium rod, 9.52 mm (0.375in) dia, 99.9% (metals basis)

Product Code. 11455300
Change view
Click to view available options
Quantity:
10 mm
50 mm
Unit Size:
10mm
50mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455300 50 mm 50mm
11465300 10 mm 10mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455300 Supplier Thermo Scientific Alfa Aesar Supplier No. 044545.DJ
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-53-1 | Eu | 151.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in mineral oil
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-53-1
Molecular Formula Eu
Molecular Weight (g/mol) 151.96
MDL Number MFCD00010992
InChI Key OGPBJKLSAFTDLK-UHFFFAOYSA-N
Synonym europio, atom, ingot, acmc-1bj8v, europium, chip, in mineral oil, foil, 1.0mm 0.04in thick, foil, 0.1mm 0.004in thick, foil, 0.25mm 0.01in thick, rod, 9.52 mm 0.375in dia, ingot reo , packed in mineral oil
PubChem CID 23981
ChEBI CHEBI:32999
IUPAC Name europium
SMILES [Eu]

Specifications

Color Gray-Silver
Quantity 50 mm
Odor Odorless
Weight ≈0.37g/mm
Form Cast Rod
UN Number UN3208
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Europium rod, ≈9.52 mm (0.375 in.) dia.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.